Learn More
Cayman Chemical ResolvIn D1 methyl ester 25UG
SDP
Catalog No. 502893189
Encompass_Preferred
A methyl ester version of the free acid that may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3189 Supplier Cayman Chemical Supplier No. 900181525 UG
A methyl ester version of the free acid that may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties