Learn More
Medchemexpress LLC Resolvin D2 | 810668-37-2 | 99.9% | 376.49 | 25 UG
SDP
Catalog No. 5000438720
Encompass_Preferred
Resolvin D2 is a metabolite derived from docosahexaenoic acid (DHA) that exhibits anti-inflammatory and anti-infective properties. It acts as a potent regulator of leukocytes and is effective in controlling microbial sepsis. This compound also functions as a remarkably potent inhibitor of TRPV1 (IC50 = 0.1 nM) and TRPA1 (IC50 = 2 nM) in primary sensory neurons.
  • Anti-inflammatory activities
  • Anti-infective activities
  • Leukocyte regulation
  • Control of microbial sepsis
  • TRPV1 and TRPA1 inhibition
  • Resolution of inflammation and tissue regeneration
  • Decreases leukocyte:endothelial interactions
  • Inhibits inflammatory pain
Catalog No. 50-004-38720 Supplier Medchemexpress LLC Supplier No. 25UGHY121636
Resolvin D2 is a metabolite derived from docosahexaenoic acid (DHA) that exhibits anti-inflammatory and anti-infective properties. It acts as a potent regulator of leukocytes and is effective in controlling microbial sepsis. This compound also functions as a remarkably potent inhibitor of TRPV1 (IC50 = 0.1 nM) and TRPA1 (IC50 = 2 nM) in primary sensory neurons.
  • Anti-inflammatory activities
  • Anti-infective activities
  • Leukocyte regulation
  • Control of microbial sepsis
  • TRPV1 and TRPA1 inhibition
  • Resolution of inflammation and tissue regeneration
  • Decreases leukocyte:endothelial interactions
  • Inhibits inflammatory pain