Learn More
Cayman Chemical ResolvIn D2 methyl ester 10UG
SDP
Catalog No. 502893897
Encompass_Preferred
A methyl ester version of RvD2 which may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3897 Supplier Cayman Chemical Supplier No. 900149710 UG
A methyl ester version of RvD2 which may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties