Learn More
Cayman Chemical ResolvIn D2 n-3 DPA 100ug
SDP
Catalog No. 502951022
Encompass_Preferred
A specialized pro-resolving mediator; formed from n-3 DPA in human leukocytes; reduces TNF-α-induced chemotaxis and adhesion of isolated human neutrophils at 1 nM; reduces peritoneal neutrophil infiltration and exudate levels of IL-6 and CCL2 in a mouse model of zymosan-induced inflammation
Catalog No. 50-295-1022 Supplier Cayman Chemical Supplier No. 34482100 UG
A specialized pro-resolving mediator; formed from n-3 DPA in human leukocytes; reduces TNF-α-induced chemotaxis and adhesion of isolated human neutrophils at 1 nM; reduces peritoneal neutrophil infiltration and exudate levels of IL-6 and CCL2 in a mouse model of zymosan-induced inflammation