Learn More
Cayman Chemical ResolvIn D2 n-3 DPA 10ug
SDP
Catalog No. 502949914
Encompass_Preferred
A specialized pro-resolving mediator; formed from n-3 DPA in human leukocytes; reduces TNF-α-induced chemotaxis and adhesion of isolated human neutrophils at 1 nM; reduces peritoneal neutrophil infiltration and exudate levels of IL-6 and CCL2 in a mouse model of zymosan-induced inflammation
Catalog No. 50-294-9914 Supplier Cayman Chemical Supplier No. 3448210 UG
A specialized pro-resolving mediator; formed from n-3 DPA in human leukocytes; reduces TNF-α-induced chemotaxis and adhesion of isolated human neutrophils at 1 nM; reduces peritoneal neutrophil infiltration and exudate levels of IL-6 and CCL2 in a mouse model of zymosan-induced inflammation