Learn More
Cayman Chemical Resolvin D2 Neurochemicals
SDP
Catalog No. 502890309
Encompass_Preferred
A product of the action of 15- and 5-LO on DHA; dampens excessive neutrophil trafficking to sites of inflammation; reduces zymosan-stimulated PMN infiltration by 70% at 10 pg per mouse; reduces PAF-stimulated leukocyte adherence and emigration at 1 nM; stimulates nitric oxide production decreases leukocyte-endothelial interactions; reduces leukocyte and PMN infiltration, decreases production of pro-inflammatory cytokines, and promotes phagocyte-mediated bacterial clearance in a mouse model of sepsis
Catalog No. 50-289-0309 Supplier Cayman Chemical Supplier No. 1000727925 UG
Add to Cart
Add to Cart
A product of the action of 15- and 5-LO on DHA; dampens excessive neutrophil trafficking to sites of inflammation; reduces zymosan-stimulated PMN infiltration by 70% at 10 pg per mouse; reduces PAF-stimulated leukocyte adherence and emigration at 1 nM; stimulates nitric oxide production decreases leukocyte-endothelial interactions; reduces leukocyte and PMN infiltration, decreases production of pro-inflammatory cytokines, and promotes phagocyte-mediated bacterial clearance in a mouse model of sepsis