Learn More
Cayman Chemical ResolvIn D3 100ug
SDP
Catalog No. 502955200
Encompass_Preferred
A DHA-derived product first identified in mouse inflammatory exudates; reduces neutrophil infiltration in vivo in both mouse peritonitis and dermal inflammation; enhances macrophage phagocytosis and efferocytosis
Catalog No. 50-295-5200 Supplier Cayman Chemical Supplier No. 13834100 UG
A DHA-derived product first identified in mouse inflammatory exudates; reduces neutrophil infiltration in vivo in both mouse peritonitis and dermal inflammation; enhances macrophage phagocytosis and efferocytosis