Learn More
Cayman Chemical ResolvIn D3 50ug
SDP
Catalog No. 502955233
Encompass_Preferred
A DHA-derived product first identified in mouse inflammatory exudates; reduces neutrophil infiltration in vivo in both mouse peritonitis and dermal inflammation; enhances macrophage phagocytosis and efferocytosis
Catalog No. 50-295-5233 Supplier Cayman Chemical Supplier No. 1383450 UG
A DHA-derived product first identified in mouse inflammatory exudates; reduces neutrophil infiltration in vivo in both mouse peritonitis and dermal inflammation; enhances macrophage phagocytosis and efferocytosis