Learn More
Cayman Chemical ResolvIn D3-d5 10ug
SDP
Catalog No. 502980959
Encompass_Preferred
An internal standard for the quantification of RvD3 by GC- or LC-MS
Catalog No. 50-298-0959 Supplier Cayman Chemical Supplier No. 1951210 UG
An internal standard for the quantification of RvD3 by GC- or LC-MS