Learn More
Cayman Chemical ResolvIn D3-d5 25ug
SDP
Catalog No. 502985369
Encompass_Preferred
An internal standard for the quantification of RvD3 by GC- or LC-MS
Catalog No. 50-298-5369 Supplier Cayman Chemical Supplier No. 1951225 UG
An internal standard for the quantification of RvD3 by GC- or LC-MS