Learn More
Cayman Chemical ResolvIn D3 methyl ester 100UG
SDP
Catalog No. 502893558
Encompass_Preferred
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3558 Supplier Cayman Chemical Supplier No. 9002686100 UG
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties