Learn More
Cayman Chemical ResolvIn D3 methyl ester 10UG
SDP
Catalog No. 502894743
Encompass_Preferred
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-4743 Supplier Cayman Chemical Supplier No. 900268610 UG
Add to Cart
Add to Cart
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties