Learn More
Cayman Chemical ResolvIn D3 methyl ester 25UG
SDP
Catalog No. 502892927
Encompass_Preferred
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-2927 Supplier Cayman Chemical Supplier No. 900268625 UG
A methyl ester form of resolvin D3; may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties