Learn More
Cayman Chemical ResolvIn D4 100ug
SDP
Catalog No. 502955126
Encompass_Preferred
A pro-resolving mediator that reduces neutrophil infiltration in vivo and promotes phagocytosis of bacteria, opsonized zymosan A, and apoptotic neutrophils by human macrophages
Catalog No. 50-295-5126 Supplier Cayman Chemical Supplier No. 13835100 UG
A pro-resolving mediator that reduces neutrophil infiltration in vivo and promotes phagocytosis of bacteria, opsonized zymosan A, and apoptotic neutrophils by human macrophages