Learn More
Medchemexpress LLC Resolvin D4 | 1025684-60-9 | 96.8% | 376.49 | 25 UG
SDP
Catalog No. 5000439235
Encompass_Preferred
Resolvin D4 (RvD4) is a specialized proresolving mediator. It can be produced in bioactive levels during *S. aureus* infection and stops neutrophil infiltration in sterile peritonitis and *S. aureus* skin infection. It enhances human macrophage and dermal fibroblast phagocytosis and efferocytosis.
  • Enhances macrophage efferocytosis of apoptotic neutrophils by 40-50%
  • Decreases the percentage of PMN by ~31% at 4 hours in *in vivo* studies
  • Modulates the severity of thrombo-inflammatory disease
  • Improves thrombus resolution
Catalog No. 50-004-39235 Supplier Medchemexpress LLC Supplier No. 25UGHY113533
Resolvin D4 (RvD4) is a specialized proresolving mediator. It can be produced in bioactive levels during *S. aureus* infection and stops neutrophil infiltration in sterile peritonitis and *S. aureus* skin infection. It enhances human macrophage and dermal fibroblast phagocytosis and efferocytosis.
  • Enhances macrophage efferocytosis of apoptotic neutrophils by 40-50%
  • Decreases the percentage of PMN by ~31% at 4 hours in *in vivo* studies
  • Modulates the severity of thrombo-inflammatory disease
  • Improves thrombus resolution