Learn More
Cayman Chemical ResolvIn D4 10ug
SDP
Catalog No. 502955022
Encompass_Preferred
A pro-resolving mediator that reduces neutrophil infiltration in vivo and promotes phagocytosis of bacteria, opsonized zymosan A, and apoptotic neutrophils by human macrophages
Catalog No. 50-295-5022 Supplier Cayman Chemical Supplier No. 1383510 UG
A pro-resolving mediator that reduces neutrophil infiltration in vivo and promotes phagocytosis of bacteria, opsonized zymosan A, and apoptotic neutrophils by human macrophages