Learn More
Cayman Chemical Resolvin E1d4 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000985410 UG
Encompass_Preferred
An internal standard for the quantification of resolvin E1 by GC- or LC-MS
Catalog No. 50-288-7992