Learn More
Cayman Chemical Resolvin E1d4 GC MS Standards
SDP
Catalog No. 502887874
Encompass_Preferred
An internal standard for the quantification of resolvin E1 by GC- or LC-MS
Catalog No. 50-288-7874 Supplier Cayman Chemical Supplier No. 1000985425 UG
An internal standard for the quantification of resolvin E1 by GC- or LC-MS