Learn More
Cayman Chemical ResolvIn E2 10ug
SDP
Catalog No. 502955009
Encompass_Preferred
An SPM; inhibits increases in inflammatory exudate neutrophil infiltration in a zymosan A-induced mouse model of peritonitis; hepatic levels are increased in mice fed normal chow, as well as in a mouse model of high-fat diet-induced NAFLD, by dietary supplementation with EPA; plasma levels are increased by dietary supplementation with fish oil containing ω-3 PUFAs in patients with chronic kidney disease or peripheral artery disease
Catalog No. 50-295-5009 Supplier Cayman Chemical Supplier No. 1382710 UG
An SPM; inhibits increases in inflammatory exudate neutrophil infiltration in a zymosan A-induced mouse model of peritonitis; hepatic levels are increased in mice fed normal chow, as well as in a mouse model of high-fat diet-induced NAFLD, by dietary supplementation with EPA; plasma levels are increased by dietary supplementation with fish oil containing ω-3 PUFAs in patients with chronic kidney disease or peripheral artery disease