Learn More
Bio Rad Laboratories RESOLVING BUFFER 1 LITER
Catalog No. NC9748157
Encompass
Resolving gel buffer is a premixed buffer used in gel casting that contains 1.5 M Tris-HCl (pH 8.8). Supplied as 1 L. (161-0798)
Catalog No. NC9748157 Supplier Bio Rad Laboratories Supplier No. 1610798
Resolving gel buffer is a premixed buffer used in gel casting that contains 1.5 M Tris-HCl (pH 8.8). Supplied as 1 L. (161-0798)