Learn More
Medchemexpress LLC Resomelagon acetate | 1809420-72-1 | 99.75% | 358.35 | 1 ML
SDP
Catalog No. 5000361460
Encompass_Preferred
Resomelagon acetate is a potent, orally active melanocortin receptor (MR) agonist targeting MC1 and MC3, suitable for research on obesity and chronic inflammation.
Catalog No. 50-003-61460 Supplier Medchemexpress LLC Supplier No. HY147301A10MM/1ML
Resomelagon acetate is a potent, orally active melanocortin receptor (MR) agonist targeting MC1 and MC3, suitable for research on obesity and chronic inflammation.