Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich RESOMER™ L 207 S, Poly(L-lactide)

Catalog No. 7699405G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-994-05G 5 g
76-994-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-994-05G Supplier Sigma Aldrich Supplier No. 7699405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biodegradable, biocompatible, and bioresorbable polymer composed of L-Lactide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.RESOMER is a registered trademark of Evonik Rohm GmbH

Specifications

CAS 33135-50-1
Quantity 5 g
Synonym Resomer™ L 207 S, PLLA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.