Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich RESOMER™ RG 750 S, Poly(D,L-lactide-co-glycolide)

Catalog No. 7697705G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-977-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-977-05G Supplier Sigma Aldrich Supplier No. 7697705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Resomer(R) RG 750 S, Poly(D,L-lactide-co-glycolide), a biodegradable and biocompatible aliphatic polyester, can be used in the development of nano and microparticles
  • It is a pharmaceutical polymer that can potentially be utilized in drug delivery and gene delivery
  • It forms a three-dimensional porous scaffold, which can be used in tissue engineering.RESOMER is a registered trademark of Evonik Rohm GmbH

Specifications

CAS 26780-50-7
Linear Formula HO[C2H4O2]n[C2H2O2]mCH3
Quantity 5 g
Synonym Resomer™ RG 750 S, PLGA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.