Learn More
GSC Go Science Crazy Resonance Apparatus
SCI_ED
Catalog No. S24658 Shop All GSC Products
Click to view available options
Resonance Apparatus
Catalog No.
S24658 Resonance Apparatus
1 options
Catalog No. S24658 Supplier GSC Go Science Crazy Supplier No. RESON01
Includes
Support stand, resonance tube, funnel tubing, graduated upright support and manual.
Requires
Tuning fork
Ideal for demonstration of standing waves. GSC International Resonance Apparatus can be used to observe resonance phenomena, speed of sound in air by exploiting standing waves, resonance effects in longitudinal waves, and resonance points for certain frequency and their relation with standing wavelength.
Permanently graduated scale on upright metal support for easy reading and years of use.
Partially fill the resonance tube with water and then sound a tuning fork of known frequency at the tubes open upper end of the Resonance Tube. The water level can be adjusted until the air in the tube resonates at the same frequency as the tuning fork which increases the volume of the sound.
Our Deluxe Resonance Apparatus includes:
  • Resonance tube
  • Funnel
  • Connecting length of tubing attached to sturdy base with clearly written graduations on board behind tube
Warranty and Services
1 year
Specifications
Product Type Resonance Apparatus
For Use With (Application) Demonstration of standing waves
Weight (English) 13 lb.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov