Learn More
Medchemexpress LLC Resorantel | 20788-07-2 | 99.8% | 10 MG
SDP
Catalog No. 5000231374
Encompass_Preferred
Resorantel is an anthelmintic small-molecule research reagent used to study parasitic infections in veterinary and parasitology research. It is supplied as a high-purity analytical standard suitable for biological assays and analytical applications.
  • Anthelmintic small-molecule for parasitology research.
  • High purity standard (99.8%).
  • Molecular formula C13H10BrNO3, molecular weight 308.13 g/mol.
  • Available as solids and DMSO solutions, including 10 mg quantities.
  • Suitable for analytical standards and biological research.
Catalog No. 50-002-31374 Supplier Medchemexpress LLC Supplier No. HY12147710MG
Resorantel is an anthelmintic small-molecule research reagent used to study parasitic infections in veterinary and parasitology research. It is supplied as a high-purity analytical standard suitable for biological assays and analytical applications.
  • Anthelmintic small-molecule for parasitology research.
  • High purity standard (99.8%).
  • Molecular formula C13H10BrNO3, molecular weight 308.13 g/mol.
  • Available as solids and DMSO solutions, including 10 mg quantities.
  • Suitable for analytical standards and biological research.