Learn More
Medchemexpress LLC Resorantel | 20788-07-2 | 99.8% | 50 MG
SDP
Catalog No. 5000231969
Encompass_Preferred
Resorantel is a brominated anthelmintic supplied as an analytical standard for research use. It is used in parasitology studies, including work on paramphistomiasis in cattle and sheep, and in investigations involving G. aegypticus. The compound is provided at high purity for laboratory and analytical applications.
  • High purity analytical standard (99.8%).
  • Brominated anthelmintic for parasitology research.
  • Molecular formula C13H10BrNO3, molecular weight 308.13 g/mol.
  • Supplied as a 50 mg vial for laboratory use.
  • Intended for research and analytical applications only, not for human or veterinary use.
Catalog No. 50-002-31969 Supplier Medchemexpress LLC Supplier No. HY12147750MG
Resorantel is a brominated anthelmintic supplied as an analytical standard for research use. It is used in parasitology studies, including work on paramphistomiasis in cattle and sheep, and in investigations involving G. aegypticus. The compound is provided at high purity for laboratory and analytical applications.
  • High purity analytical standard (99.8%).
  • Brominated anthelmintic for parasitology research.
  • Molecular formula C13H10BrNO3, molecular weight 308.13 g/mol.
  • Supplied as a 50 mg vial for laboratory use.
  • Intended for research and analytical applications only, not for human or veterinary use.