Learn More
ALADDIN SCIENTIFIC CORPORATION Resorcinol, 100g
Catalog No. 5000082519
Encompass_Preferred
Resorcinol is an aromatic alcohol useful in the synthesis of pharmaceuticals and various organic compounds as a chemical intermediate Resorcinol may be produced in bacteria from other phenolic compounds and may undergo further conversion to alicyclic products and eventual oxidation to acetyl-CoA Resorcinol cytotoxicity and cytokine release in human epidermal keratinocytes has been studied Resorcinol is also used in platicizers and diazo dyes It can be used in the qualitative determination of ketoses in the Seliwanoff test An aromatic alcohol used as a chemical intermediate
Catalog No. 50-000-82519 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. R111663100G
Resorcinol is an aromatic alcohol useful in the synthesis of pharmaceuticals and various organic compounds as a chemical intermediate Resorcinol may be produced in bacteria from other phenolic compounds and may undergo further conversion to alicyclic products and eventual oxidation to acetyl-CoA Resorcinol cytotoxicity and cytokine release in human epidermal keratinocytes has been studied Resorcinol is also used in platicizers and diazo dyes It can be used in the qualitative determination of ketoses in the Seliwanoff test An aromatic alcohol used as a chemical intermediate