Learn More
Cayman Chemical Resorcinol Monoacetate (technical grade), CAS: 102-29-4, 100 g
SDP
Catalog No. 502916091
Encompass_Preferred
A phenol acetate with antibacterial activity; increases collagen crosslinking and prevents B. subtilis growth on sponge surfaces when attached to collagen sponges at 2 or 5% (w/w); has been used as a starting material in the synthesis of αVβ3 and αVβ5 integrin dual inhibitors
Catalog No. 50-291-6091 Supplier Cayman Chemical Supplier No. 39019100 G
A phenol acetate with antibacterial activity; increases collagen crosslinking and prevents B. subtilis growth on sponge surfaces when attached to collagen sponges at 2 or 5% (w/w); has been used as a starting material in the synthesis of αVβ3 and αVβ5 integrin dual inhibitors