Learn More
Medchemexpress LLC Resorcinol monoacetate | 102-29-4 | 96.8% | 1 ML
SDP
Catalog No. 5000408631
Encompass_Preferred
Resorcinol monoacetate, also known as Acetylresorcinol, is a derivative of Resorcinol. It is primarily used in research for skin infections such as acne, seborrheic dermatitis, and tinea pedis, exhibiting antifungal activity against M. gypserim and C. albicans.
  • Derivative of resorcinol
  • Used in research of skin infections
  • Exhibits antifungal activity
Catalog No. 50-004-08631 Supplier Medchemexpress LLC Supplier No. HYB089410MM/1ML
Resorcinol monoacetate, also known as Acetylresorcinol, is a derivative of Resorcinol. It is primarily used in research for skin infections such as acne, seborrheic dermatitis, and tinea pedis, exhibiting antifungal activity against M. gypserim and C. albicans.
  • Derivative of resorcinol
  • Used in research of skin infections
  • Exhibits antifungal activity