Learn More
Medchemexpress LLC Resorcinol monoacetate | 102-29-4 | 96.8% | 100 MG
SDP
Catalog No. 5000409311
Encompass_Preferred
Resorcinol monoacetate (Acetylresorcinol) is a derivative of Resorcinol. It can be used in research of skin infections such as acne, seborrheic dermatitis and tinea pedis. It is for research use only and not sold to patients.
  • Used in research of skin infections such as acne, seborrheic dermatitis, and tinea pedis.
  • Exhibits antifungal activity against M. gypserim and C. albicans.
Catalog No. 50-004-09311 Supplier Medchemexpress LLC Supplier No. HYB0894100MG
Resorcinol monoacetate (Acetylresorcinol) is a derivative of Resorcinol. It can be used in research of skin infections such as acne, seborrheic dermatitis and tinea pedis. It is for research use only and not sold to patients.
  • Used in research of skin infections such as acne, seborrheic dermatitis, and tinea pedis.
  • Exhibits antifungal activity against M. gypserim and C. albicans.