Learn More
Medchemexpress LLC Resorcinol monoacetate | 102-29-4 | 96.8% | 500 MG
SDP
Catalog No. 5000409072
Encompass_Preferred
Resorcinol monoacetate (Acetylresorcinol) is a derivative of Resorcinol. It is used in research for skin infections like acne, seborrheic dermatitis, and tinea pedis. The compound exhibits in vitro antifungal activity against M. gypserim and C. albicans. In vivo studies indicate acute toxicity, leading to symptoms such as irritability, tremors, ataxia, salivation, lachrymation, piloerection, convulsions, and terminal depression.
  • Used in research for skin infections
  • Exhibits in vitro antifungal activity
  • Derivative of Resorcinol
  • Shows acute toxicity in vivo
Catalog No. 50-004-09072 Supplier Medchemexpress LLC Supplier No. HYB0894500MG
Resorcinol monoacetate (Acetylresorcinol) is a derivative of Resorcinol. It is used in research for skin infections like acne, seborrheic dermatitis, and tinea pedis. The compound exhibits in vitro antifungal activity against M. gypserim and C. albicans. In vivo studies indicate acute toxicity, leading to symptoms such as irritability, tremors, ataxia, salivation, lachrymation, piloerection, convulsions, and terminal depression.
  • Used in research for skin infections
  • Exhibits in vitro antifungal activity
  • Derivative of Resorcinol
  • Shows acute toxicity in vivo