Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Resorcinol monoacetate

Catalog No. R856100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R856100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R856100G Supplier Sigma Aldrich Supplier No. R856100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Resorcinol monoacetate

Specifications

CAS 102-29-4
Density 1.223 g/mL at 25°C (lit.)
Boiling Point 283°C (lit.)
Molecular Formula C8H8O3
Refractive Index n20/D 1.537 (lit.)
Linear Formula CH3CO2C6H4-3-(OH)
MDL Number MFCD00002266
Quantity 100 g
Molecular Weight (g/mol) 152.15
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.