Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol Monoacetate (Tech.), Thermo Scientific™

Catalog No. AC132305000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC132305000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC132305000 Supplier Thermo Scientific Chemicals Supplier No. 132305000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-29-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key ZZPKZRHERLGEKA-UHFFFAOYSA-N
Synonym resorcinol monoacetate, acetylresorcinol, euresol, remonol, 3-acetoxyphenol, resorcitate, m-hydroxyphenyl acetate, 1,3-benzenediol, monoacetate, resorcinol, monoacetate, resorcinol acetate
PubChem CID 5055
ChEBI CHEBI:29672
IUPAC Name (3-hydroxyphenyl) acetate
SMILES CC(=O)OC1=CC=CC(=C1)O

Specifications

Color Yellow to Brown
Infrared Spectrum Authentic
Refractive Index 1.5340 to 1.5400 (20°C, 589nm)
Quantity 500 g
Beilstein 06, 816
Merck Index 15, 8276
Formula Weight 152.15
Percent Purity ≥45% GC (≈50% resorcinol monoacetate, balance resorcinol diacetate and dihydroxybenzene)
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Resorcinol monoacetate, tech.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.