Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019687 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9687 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9687 Supplier MilliporeSigma Supelco Supplier No. PHR1201500MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1126°F
CAS 108-46-3
Vapor Pressure 1 mmHg (21.1°C)
Vapor Density 3.8 (vs. air)
Flash Point 127°C - Closed Cup
Physical Form Neat
Linear Formula C6H4-1,3-(OH)2
MDL Number MFCD00002269
Melting Point 109°C to 112°C
Boiling Point 178°C (16 mmHg)
Synonym Resorcinol; 1,3-Benzenediol
Type Certified Reference Material
Quantity 500 mg
Formula Weight 110.11
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.