Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol USP Fine Powder, Macron Fine Chemicals™

Catalog No. MK7232500 Shop All Avantor Products
Change view
Click to view available options
Glass; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-7232-500 Glass; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7232-500 Supplier Avantor Supplier No. 723203

Specifications

CAS 108-46-3
Molecular Formula C6H6O2
Packaging Glass
Quantity 500 g
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)O)O
IUPAC Name benzene-1,3-diol
Molecular Weight (g/mol) 110.112
PubChem CID 5054
ChEBI CHEBI:27810
Formula Weight 110.11
Grade USP
Physical Form Fine Powder
Chemical Name or Material Resorcinol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.