Learn More
Cayman Chemical Resorcinolnaphthalein, Purity: ≥98%, CAS: 41307-63-5, 100 mg
SDP
Catalog No. 502954160
Encompass_Preferred
Increases the activity of angiotensin-converting enzyme 2 (ACE2) in vitro (EC50 = 19.5 μM)
Catalog No. 50-295-4160 Supplier Cayman Chemical Supplier No. 13082100 MG
Increases the activity of angiotensin-converting enzyme 2 (ACE2) in vitro (EC50 = 19.5 μM)