Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resorcinolnaphthalein | 41307-63-5 | 99.9% | 10 MG
SDP

Catalog No. 5000232480
Encompass_Preferred

Resorcinolnaphthalein is a small-molecule research reagent that functions as an angiotensin-converting enzyme 2 (ACE2) enhancer. It is supplied as a solid with high reported purity and is intended for laboratory research use only.

  • CAS number 41307-63-5.
  • Molecular formula C24H14O5.
  • Molecular weight 382.36 g/mol.
  • Purity 99.9% (reported).
  • Appearance solid, light yellow to orange.
  • Biological activity ACE2 enhancer; EC50 19.5 μM.
  • Storage recommendations: powder -20°C up to 3 years, 4°C up to 2 years; in solvent -80°C up to 2 years, -20°C up to 1 year.

Catalog No. 50-002-32480 Supplier Medchemexpress LLC Supplier No. HY12244510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resorcinolnaphthalein is a small-molecule research reagent that functions as an angiotensin-converting enzyme 2 (ACE2) enhancer. It is supplied as a solid with high reported purity and is intended for laboratory research use only.

  • CAS number 41307-63-5.
  • Molecular formula C24H14O5.
  • Molecular weight 382.36 g/mol.
  • Purity 99.9% (reported).
  • Appearance solid, light yellow to orange.
  • Biological activity ACE2 enhancer; EC50 19.5 μM.
  • Storage recommendations: powder -20°C up to 3 years, 4°C up to 2 years; in solvent -80°C up to 2 years, -20°C up to 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.