Learn More
Medchemexpress LLC Resorcinolnaphthalein | 41307-63-5 | 99.9% | 50 MG
SDP
Catalog No. 5000231271
Encompass_Preferred
Resorcinolnaphthalein is a small-molecule ACE2 enhancer used for biochemical and pharmacological research. It has molecular formula C24H14O5, a molecular weight of 382.36 g/mol, and is supplied with high purity (99.87%). Intended for research use only.
  • High purity for analytical and biological studies.
  • Acts as an ACE2 enhancer for studies of ACE2-related pathways.
  • Available in multiple small research pack sizes.
  • Certificate of analysis and safety data sheet available.
  • Suitable for in vitro and in vivo research on hypertension and renal or cardiovascular models.
Catalog No. 50-002-31271 Supplier Medchemexpress LLC Supplier No. HY12244550MG
Resorcinolnaphthalein is a small-molecule ACE2 enhancer used for biochemical and pharmacological research. It has molecular formula C24H14O5, a molecular weight of 382.36 g/mol, and is supplied with high purity (99.87%). Intended for research use only.
  • High purity for analytical and biological studies.
  • Acts as an ACE2 enhancer for studies of ACE2-related pathways.
  • Available in multiple small research pack sizes.
  • Certificate of analysis and safety data sheet available.
  • Suitable for in vitro and in vivo research on hypertension and renal or cardiovascular models.