Learn More
Cayman Chemical Resorcinolnaphthalein, Purity: ≥98%, CAS: 41307-63-5, 50 mg
SDP
Catalog No. 502955895
Encompass_Preferred
Increases the activity of angiotensin-converting enzyme 2 (ACE2) in vitro (EC50 = 19.5 μM)
Catalog No. 50-295-5895 Supplier Cayman Chemical Supplier No. 1308250 MG
Increases the activity of angiotensin-converting enzyme 2 (ACE2) in vitro (EC50 = 19.5 μM)