Learn More
Abcam Resorufin, 20MG
Catalog No. 50996080
Encompass_Preferred
MW 213.19 Da. Stable red fluorophore probe. Widely used in cell labeling and Amplex Red type enzyme assays. It represents the reduced dye formed from Amplex Red reaction or resazurin reductions. Unstable in the presence of thiols such as dithiothreitol (DTT) and 2-mercaptoethanol.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-080 Supplier Abcam Supplier No. AB27430820MG
MW 213.19 Da. Stable red fluorophore probe. Widely used in cell labeling and Amplex Red type enzyme assays. It represents the reduced dye formed from Amplex Red reaction or resazurin reductions. Unstable in the presence of thiols such as dithiothreitol (DTT) and 2-mercaptoethanol.
The product is subject to the following: Abcam Restricted Use Statement