Learn More
AdipoGen Resorufin
SDP
Catalog No. 502560117
Encompass_Preferred
Chemical. CAS 635-78-9. Formula C12H7NO3. MW 213.19. Resorufin is a stable red fluorophore standard widely used in biochemical and molecular biology research. It is used in enzyme assay systems e.g. Amplex Red and as an indicator of cell viability in assays. Resorufin represents the reduced dye formed from Amplex Red reaction or resazurin reductions. One common application of resorufin is in the measurement of mitochondrial activity. When cells are metabolically active, they can reduce resazurin to resorufin, producing a measurable fluorescent signal. This property makes resorufin a valuable tool in various research applications, such as assessing cell health, drug screening, and enzyme activity studies. It is unstable in the presence of thiols such as dithiothreitol DTT and 2-mercaptoethanol. This compound can be used as a standard for enzyme substrates whose end product is resorufin.
Catalog No. 50-256-0117 Supplier AdipoGen Supplier No. CDXR0058G005
Chemical. CAS 635-78-9. Formula C12H7NO3. MW 213.19. Resorufin is a stable red fluorophore standard widely used in biochemical and molecular biology research. It is used in enzyme assay systems e.g. Amplex Red and as an indicator of cell viability in assays. Resorufin represents the reduced dye formed from Amplex Red reaction or resazurin reductions. One common application of resorufin is in the measurement of mitochondrial activity. When cells are metabolically active, they can reduce resazurin to resorufin, producing a measurable fluorescent signal. This property makes resorufin a valuable tool in various research applications, such as assessing cell health, drug screening, and enzyme activity studies. It is unstable in the presence of thiols such as dithiothreitol DTT and 2-mercaptoethanol. This compound can be used as a standard for enzyme substrates whose end product is resorufin.