Learn More
Medchemexpress LLC Resorufin | 635-78-9 | 98.9% | 213.19 g/mol | C12H7NO3 | 500MG
SDP
Catalog No. 5000238620
Encompass_Preferred
Resorufin (CAS 635-78-9) is a highly fluorescent pink dye used as a reporter for reactive oxygen and nitrogen species and as a redox/enzymatic assay indicator. It is water-soluble, exhibits red-shifted absorption and emission suitable for fluorescence-based detection and staining, and is supplied as a high-purity solid with recommended storage to protect light-sensitive samples.
  • Highly fluorescent pink dye for ROS/RNS detection.
  • Water-soluble with red-shifted absorption and emission.
  • High purity (98.9%) suitable for analytical applications.
  • Available in multiple packaging sizes, including 500 mg for larger-scale use.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-38620 Supplier Medchemexpress LLC Supplier No. HY123533500MG
Resorufin (CAS 635-78-9) is a highly fluorescent pink dye used as a reporter for reactive oxygen and nitrogen species and as a redox/enzymatic assay indicator. It is water-soluble, exhibits red-shifted absorption and emission suitable for fluorescence-based detection and staining, and is supplied as a high-purity solid with recommended storage to protect light-sensitive samples.
  • Highly fluorescent pink dye for ROS/RNS detection.
  • Water-soluble with red-shifted absorption and emission.
  • High purity (98.9%) suitable for analytical applications.
  • Available in multiple packaging sizes, including 500 mg for larger-scale use.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term stability.