Learn More
Medchemexpress LLC Resorufin | 635-78-9 | MFCD00128991 | 98.9% | 213.19 | C12H7NO3 | 5MG
SDP
Catalog No. 5000238477
Encompass_Preferred
Resorufin is a highly fluorescent pink dye used as a probe for detection of reactive oxygen species (ROS) and reactive nitrogen species (RNS) in biochemical and cell-based assays. It is water-soluble, photostable, and exhibits red-shifted optical properties, making it suitable for fluorescence measurements with excitation near 571 nm and emission near 584 nm. The compound is supplied as a solid with high chemical purity.
Catalog No. 50-002-38477 Supplier Medchemexpress LLC Supplier No. HY1235335MG
Resorufin is a highly fluorescent pink dye used as a probe for detection of reactive oxygen species (ROS) and reactive nitrogen species (RNS) in biochemical and cell-based assays. It is water-soluble, photostable, and exhibits red-shifted optical properties, making it suitable for fluorescence measurements with excitation near 571 nm and emission near 584 nm. The compound is supplied as a solid with high chemical purity.