Learn More
Medchemexpress LLC Resorufin | 635-78-9 | MFCD00128991 | 98.9% | 213.19 g/mol | C12H7NO3 | 10MG
SDP
Catalog No. 5000238728
Encompass_Preferred
Resorufin is a highly fluorescent pink dye used as a probe for detecting reactive oxygen species (ROS) and reactive nitrogen species (RNS) in biochemical and cell-based assays. It exhibits red-shifted absorption and emission and is supplied as a solid for laboratory use.
  • Highly fluorescent dye suitable for ROS and RNS detection.
  • Water-soluble with red-shifted absorption and emission properties.
  • Applicable as a fluorescent standard or probe in enzyme and cell assays.
  • Supplied as a solid with brown to black appearance for convenient storage.
  • Stable when stored protected from light; follow recommended storage conditions.
  • Available in small research quantities suitable for assay development.
Catalog No. 50-002-38728 Supplier Medchemexpress LLC Supplier No. HY12353310MG
Resorufin is a highly fluorescent pink dye used as a probe for detecting reactive oxygen species (ROS) and reactive nitrogen species (RNS) in biochemical and cell-based assays. It exhibits red-shifted absorption and emission and is supplied as a solid for laboratory use.
  • Highly fluorescent dye suitable for ROS and RNS detection.
  • Water-soluble with red-shifted absorption and emission properties.
  • Applicable as a fluorescent standard or probe in enzyme and cell assays.
  • Supplied as a solid with brown to black appearance for convenient storage.
  • Stable when stored protected from light; follow recommended storage conditions.
  • Available in small research quantities suitable for assay development.