Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resorufin | 635-78-9 | MFCD00128991 | 98.9% | 213.19 g/mol | C12H7NO3 | 1G
SDP

Catalog No. 5000236423
Encompass_Preferred

Resorufin is a highly fluorescent pink dye used as a fluorometric reporter for detecting reactive oxygen species (ROS) and reactive nitrogen species (RNS). It is water-soluble and exhibits red-shifted excitation/emission, making it suitable as a standard or indicator in fluorescence-based assays.

  • Highly fluorescent pink dye for ROS/RNS detection.
  • Excitation 571 nm; emission 584 nm.
  • Molecular formula C12H7NO3; molecular weight 213.19 g/mol.
  • Purity 98.9% (as provided).
  • Solid, brown to black; protect from light during storage.
  • Storage guidance: 4°C short term; in solvent -80°C for 6 months or -20°C for 1 month.
  • Available in 1 g packaging suitable for research use.

Catalog No. 50-002-36423 Supplier Medchemexpress LLC Supplier No. HY1235331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resorufin is a highly fluorescent pink dye used as a fluorometric reporter for detecting reactive oxygen species (ROS) and reactive nitrogen species (RNS). It is water-soluble and exhibits red-shifted excitation/emission, making it suitable as a standard or indicator in fluorescence-based assays.

  • Highly fluorescent pink dye for ROS/RNS detection.
  • Excitation 571 nm; emission 584 nm.
  • Molecular formula C12H7NO3; molecular weight 213.19 g/mol.
  • Purity 98.9% (as provided).
  • Solid, brown to black; protect from light during storage.
  • Storage guidance: 4°C short term; in solvent -80°C for 6 months or -20°C for 1 month.
  • Available in 1 g packaging suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.