Learn More
Cayman Chemical Resorufin Acetate, Purity: ≥98%, CAS: 1152-14-3, 10 mg
SDP
Catalog No. 502986071
Encompass_Preferred
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin
Catalog No. 50-298-6071 Supplier Cayman Chemical Supplier No. 2168610 MG
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin