Learn More
Cayman Chemical ResorufIn Acetate 25mg
SDP
Catalog No. 502986090
Encompass_Preferred
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin
Catalog No. 50-298-6090 Supplier Cayman Chemical Supplier No. 2168625 MG
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin