Learn More
Abcam Resorufin acetate, 25MG
Catalog No. 50996141
Encompass_Preferred
MW 255.22 Da, Purity >98%. Red fluorogenic esterase substrate, useful in cell viability assays, or for high throughput screening of esterase activity in a live cell format.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-141 Supplier Abcam Supplier No. AB27487725MG
MW 255.22 Da, Purity >98%. Red fluorogenic esterase substrate, useful in cell viability assays, or for high throughput screening of esterase activity in a live cell format.
The product is subject to the following: Abcam Restricted Use Statement