Learn More
Cayman Chemical ResorufIn Acetate 50mg
SDP
Catalog No. 502963987
Encompass_Preferred
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin
Catalog No. 50-296-3987 Supplier Cayman Chemical Supplier No. 2168650 MG
A fluorometric probe that acts as a substrate for ALDH1A1 esterase activity and chymotrypsin