Learn More
AdipoGen Resorufin-b-D-galactopyranosid
SDP
Catalog No. 502560153
Encompass_Preferred
Chemical. CAS 95079-19-9. Formula C18H17NO8. MW BD9837. Resorufin beta-D-galactopyranoside is a fluorogenic substrate for beta-galactosidase. Upon enzymatic cleavage by the enzymes, the fluorescent hydrolysis product resorufin is released and its fluorescence can be used to quantify enzyme activity. Spectral data of Resorufin lambdaex=571nm, lambdaem=584nm. It is used in several applications, specifically in compound screens, where it is advantageous because it is less prone to compound autofluorescence, and thus produces less false positives. It is widely used as substrate in high throughput screening of galactosidase inhibitors.
Catalog No. 50-256-0153 Supplier AdipoGen Supplier No. CDXR0005M010
Chemical. CAS 95079-19-9. Formula C18H17NO8. MW BD9837. Resorufin beta-D-galactopyranoside is a fluorogenic substrate for beta-galactosidase. Upon enzymatic cleavage by the enzymes, the fluorescent hydrolysis product resorufin is released and its fluorescence can be used to quantify enzyme activity. Spectral data of Resorufin lambdaex=571nm, lambdaem=584nm. It is used in several applications, specifically in compound screens, where it is advantageous because it is less prone to compound autofluorescence, and thus produces less false positives. It is widely used as substrate in high throughput screening of galactosidase inhibitors.